α-Chloro-3,4-difluorotoulene 98%

Supplier: Thermo Scientific Chemicals
Danger

38442-0050 38442-0250
TS38442-0050EA 77 USD
TS38442-0050 TS38442-0250
α-Chloro-3,4-difluorotoulene 98%
α-Chloro-3,4-difluorotoulene
Formula: C₇H₅ClF₂
MDL Number: MFCD00236218
CAS Number: 698-80-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4940 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR