4-Chloro-m-toluidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-3-methylaniline, 5-Amino-2-chlorotoluene

38368-0050 38368-0250
TS38368-0050EA 55 USD
TS38368-0050 TS38368-0250
4-Chloro-m-toluidine 98+%
4-Chloro-m-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 83…84 °C
MDL Number: MFCD00066332
CAS Number: 7149-75-9
UN: 2239
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 83 to 84 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR