2-Bromobiphenyl (PBB No. 1) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromobiphenyl, PBB No. 1, 2-Bromodiphenyl

38345-0050 38345-0250
TS38345-0010EA 30.9 USD
TS38345-0010 TS38345-0050 TS38345-0250
2-Bromobiphenyl (PBB No. 1) 98%
2-Bromobiphenyl (PBB No. 1)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 297…298 °C (760 mmHg)
Melting Pt: 1.5…2 °C
Density: 1.352 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000065
CAS Number: 2052-07-5
EINECS: 218-141-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.6280 to 1.6300 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR