1-(2-Methylphenyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

38213-0010 38213-0050
TS38213-0010EA 33 USD
TS38213-0010 TS38213-0050
1-(2-Methylphenyl)piperazine 98%
1-(2-Methylphenyl)piperazine
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
Melting Pt: 48…52 °C
MDL Number: MFCD00040729
CAS Number: 39512-51-1
EINECS: 254-481-9
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 43 to 48 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR