1-Fluoro-2-ethynylbenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Fluorophenyl)ethyne, 1-Ethynyl-2-fluorobenzene, (2-Fluorophenyl)acetylene

38099-0010 38099-2500
TS38099-0010EA 58 USD
TS38099-0010 TS38099-2500
1-Fluoro-2-ethynylbenzene 97%
1-Fluoro-2-ethynylbenzene
Formula: C₈H₅F
MW: 120.13 g/mol
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799540
CAS Number: 766-49-4
UN: 1993

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5270 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR