1-Bromo-4-fluoro-3-nitrobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
38089-0050
38089-0250
TS38089-0050EA
70
USD
TS38089-0050
TS38089-0250
1-Bromo-4-fluoro-3-nitrobenzene 98%
1-Bromo-4-fluoro-3-nitrobenzene
Formula:
C₆H₃BrFNO₂ Boiling Pt: 145…147 °C (35 mmHg) Melting Pt: 18…20 °C Density: 1.786 g/cm³ Flash Pt: 110 °C |
MDL Number:
MFCD00129165 CAS Number: 364-73-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow to brown Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5750 to 1.5780 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...