Benzyl propargyl ether 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ((Prop-2-yn-1-yloxy)methyl)benzene

38087-0010 38087-0050
TS38087-0010EA 31 USD
TS38087-0010 TS38087-0050
Benzyl propargyl ether 97%
Benzyl propargyl ether
Formula: C₁₀H₁₀O
MW: 146.189 g/mol
Boiling Pt: 100…101 °C (18 mmHg)
Density: 1.011 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00483987
CAS Number: 4039-82-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5220 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR