2-Bromophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Bromophenyl)boronic acid, Dihydroxy(2-bromophenyl)borane, 2-Bromobenzeneboronic acid, (2-Bromophenyl)metaboric acid

37999-0050 37999-0250 37999-1000
TS37999-0050EA 91 USD
TS37999-0050 TS37999-0250 TS37999-1000
2-Bromophenylboronic acid 97%
2-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
Melting Pt: 109…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01114672
CAS Number: 244205-40-1

Order Now

Specification Test Results

Appearance White to light yellow Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR