(2,6-Difluorophenyl)acetone 98%

Supplier: Thermo Scientific Chemicals

37915-0010 37915-0050
TS37915-0010EA 68 USD
TS37915-0010 TS37915-0050
(2,6-Difluorophenyl)acetone 98%
(2,6-Difluorophenyl)acetone
Formula: C₉H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD02258867
CAS Number: 101712-20-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4820 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR