4-Ethylbenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Ethylbenzaldehyde

37895-0050 37895-0250 37895-1000
TS37895-0050EA 21 USD
TS37895-0050 TS37895-0250 TS37895-1000
4-Ethylbenzaldehyde 98%
4-Ethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 221 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006956
CAS Number: 4748-78-1
EINECS: 225-268-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR