2-Ethylbenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37875-0010
37875-0050
TS37875-0010EA
180
USD
TS37875-0010
TS37875-0050
2-Ethylbenzoic acid 97%
2-Ethylbenzoic acid
Formula:
C₉H₁₀O₂ Melting Pt: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045838 CAS Number: 612-19-1 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 62 to 66 °C |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...