(R)-(-)-2-Phenylbutyric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(-)-2-Phenylbutanoic acid

37852-0010 37852-0050
TS37852-0010EA 132 USD
TS37852-0010 TS37852-0050
(R)-(-)-2-Phenylbutyric acid 99%
(R)-(-)-2-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 130 °C (3mbar)
Density: 1.055 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00063165
CAS Number: 938-79-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5170 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR