1-Chloro-4-ethynylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne

37706-0010 37706-2500
TS37706-0010EA 151 USD
TS37706-0010 TS37706-2500
1-Chloro-4-ethynylbenzene 98%
1-Chloro-4-ethynylbenzene
Formula: C₈H₅Cl
Boiling Pt: 79…82 °C (23 mmHg)
Melting Pt: 43…47 °C
Flash Pt: 10 °C
MDL Number: MFCD00191917
CAS Number: 873-73-4
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline mass
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR