(4-Fluorophenyl)acetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluorobenzeneacetyl chloride, 4-Fluorophenylacetyl Chloride

37612-0010 37612-0050 37612-0250
TS37612-0010EA 45 USD
TS37612-0010 TS37612-0050 TS37612-0250
(4-Fluorophenyl)acetyl chloride 98%
(4-Fluorophenyl)acetyl chloride
Formula: (C₆H₄F)CH₂COCl
MW: 172.59 g/mol
Boiling Pt: 60 °C (8 mmHg)
Density: 1.259 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00674181
CAS Number: 459-04-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5130 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR