(2,4-Difluorophenyl)acetone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2,4-Difluorophenyl)propan-2-one

37608-0010 37608-0050
TS37608-0010EA 37 USD
TS37608-0010 TS37608-0050
(2,4-Difluorophenyl)acetone 98%
(2,4-Difluorophenyl)acetone
Formula: C₉H₈F₂O
MW: 170.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258866
CAS Number: 274682-91-6

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4790 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR