2-(Benzyloxy)acetaldehyde 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Benzyloxy)acetaldehyde, Benzyloxyacetaldehyde

37607-0010 37607-0050
TS37607-0010EA 57 USD
TS37607-0010 TS37607-0050
2-(Benzyloxy)acetaldehyde 95% stabilized
2-(Benzyloxy)acetaldehyde
Formula: C₉H₁₀O₂
MW: 150.177 g/mol
Boiling Pt: 118…120 °C (13 mmHg)
Density: 1.069 g/cm³
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191779
CAS Number: 60656-87-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
Stabilizer 0.5 % hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR