3-Fluoro-p-anisidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Fluoro-4-anisidine, 3-Fluoro-4-methoxyaniline, 4-Amino-2-fluoroanisole
37588-0010
37588-0050
TS37588-0010EA
37
USD
TS37588-0010
TS37588-0050
3-Fluoro-p-anisidine 99%
3-Fluoro-p-anisidine
Formula:
C₇H₈FNO MW: 141.15 g/mol Boiling Pt: 135 °C (18 mmHg) Melting Pt: 81…84 °C |
MDL Number:
MFCD00075040 CAS Number: 366-99-4 |
Specification Test Results
Appearance | Light beige to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 81 to 84 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...