4-(Trifluoromethyl)benzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

37583-0010 37583-0100
TS37583-0010EA 31 USD
TS37583-0010 TS37583-0100
4-(Trifluoromethyl)benzylamine 97%
4-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 79…82 °C (15 mmHg)
Density: 1.229 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010220
CAS Number: 3300-51-4
EINECS: 221-971-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4650 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR