4-Mercaptobenzoic acid 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Thiobenzoic acid

37330-0050 37330-0250
TS37330-0050EA 88 USD
TS37330-0050 TS37330-0250
4-Mercaptobenzoic acid 90%
4-Mercaptobenzoic acid
Formula: C₇H₆O₂S
MW: 154.1863 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00016617
CAS Number: 1074-36-8
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 215 to 224 °C
HPLC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR