3-Phenylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

37177-0010 37177-0050
TS37177-0010EA 27 USD
TS37177-0010 TS37177-0050
3-Phenylbenzoic acid 97%
3-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00045846
CAS Number: 716-76-7

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR