(R)-(+)-4-Chloro-α-methylbenzylamine 97%, 98% ee
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(+)-1-(4-Chlorophenyl)ethylamine, (R)-1-(4-Chlorophenyl)ethanamine, (R)-1-(4-Chlorophenyl)ethylamine
37065-0010
37065-0050
37065-0250
TS37065-0010EA
38
USD
TS37065-0010
TS37065-0050
TS37065-0250
(R)-(+)-4-Chloro-α-methylbenzylamine 97%, 98% ee
(R)-(+)-4-Chloro-α-methylbenzylamine
Formula:
C₈H₁₀ClN MW: 155.63 g/mol Boiling Pt: 232 °C (760 mmHg) Density: 1.115 g/cm³ Flash Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671639 CAS Number: 27298-99-3 UN: 2810 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless to pale yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5400 to 1.5440 (20°C, 589 nm) |
GC | ≥96.0 % |
Enantiomeric excess | ≥97.5 % |
Specific optical rotation | +29° to +33° (20°C, 589 nm) (Neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...