2,4-Dimethoxybenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

36995-0050 36995-0250
TS36995-0050EA 57 USD
TS36995-0050 TS36995-0250
2,4-Dimethoxybenzylamine 98%
2,4-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
Boiling Pt: 140 °C (760 mmHg)
Density: 1.113 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052393
CAS Number: 20781-20-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR