Methyl-3-(bromomethyl)benzoate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 3-(bromomethyl)benzoate

36863-0050 36863-0250 36863-1000
TS36863-0050EA 97 USD
TS36863-0050 TS36863-0250 TS36863-1000
Methyl-3-(bromomethyl)benzoate 95%
Methyl 3-(bromomethyl)benzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 135 °C (2 mmHg)
Melting Pt: 35…37 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051437
CAS Number: 1129-28-8
EINECS: 419-100-4
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 47 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR