3-Iodobenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

36860-0010 36860-0100
TS36860-0010EA 15 USD
TS36860-0010 TS36860-0100
3-Iodobenzonitrile 97%
3-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Melting Pt: 39…44 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00079762
CAS Number: 69113-59-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 38 to 44 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR