Benzophenone imine 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzophenoneimine

36859-0050 36859-0250 36859-1000
TS36859-0050EA 68 USD
TS36859-0050 TS36859-0250 TS36859-1000
Benzophenone imine 97% stabilized
Benzophenone imine
Formula: C₁₃H₁₁N
MW: 181.24 g/mol
Boiling Pt: 216 °C (75 mmHg)
Melting Pt: –30 °C
Density: 1.08 g/cm³
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00001760
CAS Number: 1013-88-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6170 to 1.6190 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Approx. 0.02 % w/w Hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR