(4-Acetamidophenyl)boronic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Acetamidophenylboronic acid, 4-Acetamidobenzeneboronic acid, 4-Acetylaminophenylboronic acid
36761-0010
36761-0050
TS36761-0010EA
59
USD
TS36761-0010
TS36761-0050
(4-Acetamidophenyl)boronic acid 97+%
(4-Acetamidophenyl)boronic acid
Formula:
C₈H₁₀BNO₃ MW: 178.98 g/mol Melting Pt: 215…220 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179451 CAS Number: 101251-09-6 |
Specification Test Results
Appearance | Grey to tan Powder |
Infrared spectrum | Conforms |
GC | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...