2,4-Dimethylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

36553-0010 36553-0050
TS36553-0010EA 65 USD
TS36553-0010 TS36553-0050
2,4-Dimethylbenzylamine 98%
2,4-Dimethylbenzylamine
Formula: C₉H₁₃N
Boiling Pt: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00025575
CAS Number: 94-98-4
EINECS: 202-380-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5390 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR