Methyl-6-bromo-2-naphthoate 98%

Supplier: Thermo Scientific Chemicals

36414-0050 36414-0250 36414-1000
TS36414-0050EA 39 USD
TS36414-0050 TS36414-0250 TS36414-1000
Methyl-6-bromo-2-naphthoate 98%
Methyl 6-bromo-2-naphthoate
Formula: C₁₂H₉BrO₂
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00100408
CAS Number: 33626-98-1

Order Now

Specification Test Results

Appearance Off-white to cream to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 126 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR