4-Fluoro-L-2-phenylglycine ≥95%, ee 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Fluoro-L-phenylglycine, (S)-2-Amino-2-(4-fluorophenyl)acetic Acid, H-Phg(4-F)-OH, (S)-4-Fluorophenylglycine
36205-0010
36205-0050
TS36205-0010EA
202
USD
TS36205-0010
TS36205-0050
4-Fluoro-L-2-phenylglycine ≥95%, ee 98%
4-Fluoro-L-2-phenylglycine
Formula:
C₈H₈FNO₂ MW: 169.16 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213799 CAS Number: 19883-57-9 |
Specification Test Results
Appearance | Off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Enantiomeric excess | ≥97.5 % (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...