(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-Ethyl 3-amino-3-phenylpropanoate hydrochloride

36204-0010 36204-0250
TS36204-0010EA 114 USD
TS36204-0010 TS36204-0250
(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
Melting Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD02259715
CAS Number: 167834-24-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR