4-(Bromomethyl)benzenesulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)benzene-1-sulphonyl chloride

36054-0010 36054-0050 36054-0250
TS36054-0010EA 30 USD
TS36054-0010 TS36054-0050 TS36054-0250
4-(Bromomethyl)benzenesulfonyl chloride 95%
4-(Bromomethyl)benzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 69…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00156129
CAS Number: 66176-39-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 72 to 77 °C
Titration Argentometric ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR