4-Phenoxyphenylboronic acid 95+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

35964-0010 35964-0050 35964-0250
TS35964-0010EA 34 USD
TS35964-0010 TS35964-0050 TS35964-0250
4-Phenoxyphenylboronic acid 95+%
4-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00093312
CAS Number: 51067-38-0

Order Now

Specification Test Results

Appearance White to off-white Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR