4-(Methylsulfonyl)phenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

35951-0010 35951-0050
TS35951-0010EA 73 USD
TS35951-0010 TS35951-0050
4-(Methylsulfonyl)phenylboronic acid 98+%
4-(Methylsulphonyl)phenylboronic acid
Formula: C₇H₉BO₄S
MW: 200.02 g/mol
Melting Pt: 275…277 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01630820
CAS Number: 149104-88-1

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR