4-Formylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

35938-0010 35938-0050 35938-0250
TS35938-0010EA 34 USD
TS35938-0010 TS35938-0050 TS35938-0250
4-Formylphenylboronic acid 97%
4-Formylphenylboronic acid
Formula: C₇H₇BO₃
Melting Pt: 256…257 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151823
CAS Number: 87199-17-5

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR