2-Formylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane

35936-0010 35936-0050
TS35936-0010EA 48 USD
TS35936-0010 TS35936-0050
2-Formylphenylboronic acid 98%
2-Formylphenylboronic acid
Formula: C₇H₇BO₃
Melting Pt: 120…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151822
CAS Number: 40138-16-7

Order Now

Specification Test Results

Appearance White to pink to orange Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR