2,6-Dimethylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dimethylbenzeneboronic acid, (2,6-Dimethylphenyl)boronic acid, Dihydroxy(2,6-dimethylphenyl)borane, (2,6-Dimethylphenyl)metaboric acid

35927-0010 35927-0050 35927-0250
TS35927-0010EA 34 USD
TS35927-0010 TS35927-0050 TS35927-0250
2,6-Dimethylphenylboronic acid 98%
2,6-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 105 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR