3-Chlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane

35900-0010 35900-0050
TS35900-0010EA 25 USD
TS35900-0010 TS35900-0050
3-Chlorophenylboronic acid 97%
3-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00161354
CAS Number: 63503-60-6

Order Now

Specification Test Results

Appearance White to yellow-green Crystalline powder or needles
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR