2-Chlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid

35899-0010 35899-0050
TS35899-0010EA 38 USD
TS35899-0010 TS35899-0050
2-Chlorophenylboronic acid 97%
2-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
Melting Pt: 94…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674012
CAS Number: 3900-89-8

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals or granules
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR