4-tert-Butylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid

35897-0050 35897-0250
TS35897-0050EA 81 USD
TS35897-0050 TS35897-0250
4-tert-Butylphenylboronic acid 97%
4-tert-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
Melting Pt: 191…196 °C
Storage Temperature: Ambient
MDL Number: MFCD01009697
CAS Number: 123324-71-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR