4-Iodo-m-toluidine 98%

Supplier: Thermo Scientific Chemicals
Warning

35780-0050 35780-0250 35780-1000
TS35780-0050EA 49 USD
TS35780-0050 TS35780-0250 TS35780-1000
4-Iodo-m-toluidine 98%
4-Iodo-m-toluidine
Formula: C₇H₈IN
Melting Pt: 50…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01569451
CAS Number: 4949-69-3

Order Now

Specification Test Results

Appearance Beige to light brown to dark grey Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 55 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR