4-(Methylsulphonyl)benzyl bromide tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α-Bromo-4-(methylsulphonyl)toluene, 1-(Bromomethyl)-4-(methylsulphonyl)benzene
35758-0025
35758-0100
TS35758-0025EA
124
USD
TS35758-0025
TS35758-0100
4-(Methylsulphonyl)benzyl bromide tech.
4-(Methylsulphonyl)benzyl bromide
Formula:
C₈H₉BrO₂S Melting Pt: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185849 CAS Number: 53606-06-7 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥60 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...