2,2'-Dibromobiphenyl (PBB No. 4) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2-(2-bromophenyl)benzene, 2,2'-Dibromobiphenyl
35593-0010
35593-0050
TS35593-0010EA
59
USD
TS35593-0010
TS35593-0050
2,2'-Dibromobiphenyl (PBB No. 4) 98%
2,2'-Dibromobiphenyl (PBB No. 4)
Formula:
C₁₂H₈Br₂ MW: 312 g/mol Melting Pt: 79…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093707 CAS Number: 13029-09-9 UN: 3152 ADR: 9,II |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 79 to 83 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...