2-Chloro-6-fluoroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

35582-0010 35582-0050
TS35582-0010EA 68 USD
TS35582-0010 TS35582-0050
2-Chloro-6-fluoroaniline 99%
2-Chloro-6-fluoroaniline
Formula: C₆H₅ClFN
Boiling Pt: 67…69 °C (14 mmHg)
Melting Pt: 31…32 °C
Density: 1.316 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00040309
CAS Number: 363-51-9
EINECS: 206-657-1
UN: 2019
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5540 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR