3,5-Dimethylbenzyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromomesitylene, 1-(Bromomethyl)-3,5-dimethylbenzene

35556-0050 35556-0250
TS35556-0050EA 54 USD
TS35556-0050 TS35556-0250
3,5-Dimethylbenzyl bromide 98%
3,5-Dimethylbenzyl bromide
Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 101…103 °C (8 mmHg)
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00013539
CAS Number: 27129-86-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow Low melting crystalline mass
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR