(Methylsulfonyl)benzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl phenyl sulphone

35304-0050 35304-0250
TS35304-0050EA 58 USD
TS35304-0050 TS35304-0250
(Methylsulfonyl)benzene 99%
(Methylsulphonyl)benzene
Formula: C₇H₈O₂S
MW: 156.21 g/mol
Melting Pt: 85…88 °C
MDL Number: MFCD00014741
CAS Number: 3112-85-4
EINECS: 221-476-8

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 85 to 88 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR