4-Phenoxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35272-0010
35272-0050
35272-0250
TS35272-0010EA
19
USD
TS35272-0010
TS35272-0050
TS35272-0250
4-Phenoxybenzoic acid 98%
4-Phenoxybenzoic acid
Formula:
C₁₃H₁₀O₃ Melting Pt: 160…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002539 CAS Number: 2215-77-2 EINECS: 218-682-5 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 160 to 164 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...