1,2,4,5-Tetrabromobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

35240-0250 35240-1000
TS35240-0250EA 74 USD
TS35240-0250 TS35240-1000
1,2,4,5-Tetrabromobenzene 97%
1,2,4,5-Tetrabromobenzene
Formula: C₆H₂Br₄
MW: 393.7 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00000063
CAS Number: 636-28-2
EINECS: 211-253-3

Order Now

Specification Test Results

Appearance White to beige to brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 177 to 183 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR