Monomethyl terephthalate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35234-0050
35234-0250
35234-1000
TS35234-0050EA
49
USD
TS35234-0050
TS35234-0250
TS35234-1000
Monomethyl terephthalate 97%
Monomethyl terephthalate
Formula:
C₉H₈O₄ Melting Pt: 219…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002557 CAS Number: 1679-64-7 EINECS: 216-849-7 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 219 to 224 °C |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...