4-Methoxybenzaldehyde dimethyl acetal 98%

Supplier: Thermo Scientific Chemicals

Synonyms: p-(Dimethoxymethyl)anisole, P-anisaldehyde dimethyl acetal, p-Anisaldehyde dimethyl acetal, 1-(Dimethoxymethyl)-4-methoxybenzene, 4-Methoxy-α,α-dimethoxytoluene

35229-0250 35229-1000 35229-5000
TS35229-0250EA 68 USD
TS35229-0250 TS35229-1000 TS35229-5000
4-Methoxybenzaldehyde dimethyl acetal 98%
4-Methoxybenzaldehyde dimethyl acetal
Formula: C₁₀H₁₄O₃
Boiling Pt: 235 °C (760 mmHg)
Density: 1.068 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036507
CAS Number: 2186-92-7
EINECS: 218-577-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5035 to 1.5075 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR