Methyl-3-hydroxybenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester
35200-0250
35200-1000
TS35200-0250EA
68
USD
TS35200-0250
TS35200-1000
Methyl-3-hydroxybenzoate 98%
Methyl-3-hydroxybenzoate
Formula:
C₈H₈O₃ Boiling Pt: 280…281 °C (709 mmHg) Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002295 CAS Number: 19438-10-9 EINECS: 243-071-5 |
Specification Test Results
Appearance | White to beige Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 70 to 74 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...